2'-Chloro-5'-methoxy-[1,1'-biphenyl]-4-carbaldehyde ≥97%

Supplier: AOBChem USA

Warning

69075-500MG 69075-1G 69075-5G 69075-25G
77333-412EA 122.57 USD
77333-412 77344-006 77369-426 77381-514
2'-Chloro-5'-methoxy-[1,1'-biphenyl]-4-carbaldehyde ≥97%
2'-Chloro-5'-methoxy-[1,1'-biphenyl]-4-carbaldehyde

Precaución: for Research Use Only

Formula: C₁₄H₁₁ClO₂
MW: 246.689 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34642824
CAS Number: 2901099-26-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR