2-Chloro-5-methoxy-4'-(trifluoromethyl)-1,1'-biphenyl ≥95%
Supplier: AOBChem USA
|
Warning
|
69073-500MG
69073-1G
69073-5G
69073-25G
77333-410EA
101.97
USD
77333-410
77342-972
77369-424
77381-512
2-Chloro-5-methoxy-4'-(trifluoromethyl)-1,1'-biphenyl ≥95%
2-Chloro-5-methoxy-4'-(trifluoromethyl)-1,1'-biphenyl
Precaución: for Research Use Only
Formula:
C₁₄H₁₀ClF₃O MW: 286.677 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD18423965 CAS Number: 1261599-16-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...