2-Chloro-5-methoxy-4'-methyl-1,1'-biphenyl ≥97%

Supplier: AOBChem USA

Warning

69070-500MG 69070-1G 69070-5G 69070-25G
77333-406EA 95.79 USD
77333-406 77342-628 77369-420 77381-508
2-Chloro-5-methoxy-4'-methyl-1,1'-biphenyl ≥97%
2-Chloro-5-methoxy-4'-methyl-1,1'-biphenyl

Precaución: for Research Use Only

Formula: C₁₄H₁₃ClO
MW: 232.705 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34763764
CAS Number: 2901085-41-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR