2-Chloro-5-methoxy-1,1'-biphenyl ≥95%

Supplier: AOBChem USA

Warning

69069-500MG 69069-1G 69069-5G 69069-25G
77333-404EA 101.97 USD
77333-404 77342-970 77369-418 77381-506
2-Chloro-5-methoxy-1,1'-biphenyl ≥95%
2-Chloro-5-methoxy-1,1'-biphenyl

Precaución: for Research Use Only

Formula: C₁₃H₁₁ClO
MW: 218.679 g/mol
Storage Temperature: Ambient
MDL Number: MFCD14701647
CAS Number: 21345-12-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR