(2-Methyl-[1,1'-biphenyl]-3-yl)boronic acid ≥97%

Supplier: AOBChem USA

Warning

69066-500MG 69066-1G 69066-5G
77333-402EA 347.11 USD
77333-402 77357-566 77381-504
(2-Methyl-[1,1'-biphenyl]-3-yl)boronic acid ≥97%
(2-Methyl-[1,1'-biphenyl]-3-yl)boronic acid

Precaución: for Research Use Only

Formula: C₁₃H₁₃BO₂
MW: 212.052 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34471089
CAS Number: 1899060-83-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR