5'-Chloro-2'-fluoro-[1,1'-biphenyl]-4-carbaldehyde ≥95%

Supplier: AOBChem USA

Warning

69064-500MG 69064-1G 69064-5G 69064-25G
77333-400EA 223.51 USD
77333-400 77357-564 77369-416 77390-434
5'-Chloro-2'-fluoro-[1,1'-biphenyl]-4-carbaldehyde ≥95%
5'-Chloro-2'-fluoro-[1,1'-biphenyl]-4-carbaldehyde

Precaución: for Research Use Only

Formula: C₁₃H₈ClFO
MW: 234.653 g/mol
Storage Temperature: Ambient
MDL Number: MFCD26628992
CAS Number: 1540317-58-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR