(3-Methoxy-2-propoxyphenyl)methanol ≥97%

Supplier: AOBChem USA

Warning

69043-500MG 69043-1G 69043-5G 69043-25G
77333-396EA 141.11 USD
77333-396 77344-900 77369-412 77390-432
(3-Methoxy-2-propoxyphenyl)methanol ≥97%
(3-Methoxy-2-propoxyphenyl)methanol

Precaución: for Research Use Only

Formula: C₁₁H₁₆O₃
Storage Temperature: Ambient
MDL Number: MFCD12177223
CAS Number: 103862-01-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR