1-(3,5-Dichloro-4-propoxyphenyl)ethanone ≥95%

Supplier: AOBChem USA

Warning

69039-500MG 69039-1G 69039-25G 69039-5G
77333-394EA 301.79 USD
77333-394 77357-562 77390-430 77381-500
1-(3,5-Dichloro-4-propoxyphenyl)ethanone ≥95%
1-(3,5-Dichloro-4-propoxyphenyl)ethanone

Precaución: for Research Use Only

Formula: C₁₁H₁₂Cl₂O₂
Storage Temperature: Ambient
MDL Number: MFCD34763756
CAS Number: 1353220-44-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR