(4-Butoxy-3,5-dichlorophenyl)(methyl)sulfane ≥97%

Supplier: AOBChem USA

Warning

69035-500MG 69035-1G 69035-5G 69035-25G
77333-390EA 254.41 USD
77333-390 77357-560 77369-408 77390-426
(4-Butoxy-3,5-dichlorophenyl)(methyl)sulfane ≥97%
(4-Butoxy-3,5-dichlorophenyl)(methyl)sulfane

Precaución: for Research Use Only

Formula: C₁₁H₁₄Cl₂OS
Storage Temperature: Ambient
MDL Number: MFCD34763753
CAS Number: 2901102-24-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR