1-(3,5-Dichloro-4-ethoxyphenyl)ethanone ≥97%

Supplier: AOBChem USA

Warning

69032-500MG 69032-1G 69032-5G 69032-25G
77333-384EA 260.59 USD
77333-384 77357-558 77369-402 77390-420
1-(3,5-Dichloro-4-ethoxyphenyl)ethanone ≥97%
1-(3,5-Dichloro-4-ethoxyphenyl)ethanone

Precaución: for Research Use Only

Formula: C₁₀H₁₀Cl₂O₂
Storage Temperature: Ambient
MDL Number: MFCD10485812
CAS Number: 932944-55-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR