2-Fluoro-4-iodo-3-isopropoxybenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

69028-500MG 69028-1G 69028-5G 69028-25G
77333-380EA 184.37 USD
77333-380 77347-174 77369-398 77390-416
2-Fluoro-4-iodo-3-isopropoxybenzaldehyde ≥97%
2-Fluoro-4-iodo-3-isopropoxybenzaldehyde

Precaución: for Research Use Only

Formula: C₁₀H₁₀FIO₂
MW: 308.088 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34763747
CAS Number: 2918831-94-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR