4-(4-Methylbenzyl)benzoic acid ≥97%

Supplier: AOBChem USA

Warning

69023-500MG 69023-1G 69023-5G
77336-562EA 388.31 USD
77336-562 77357-554 77381-498
4-(4-Methylbenzyl)benzoic acid ≥97%
4-(4-Methylbenzyl)benzoic acid

Precaución: for Research Use Only

Formula: C₁₅H₁₄O₂
Storage Temperature: Ambient
MDL Number: MFCD32219389
CAS Number: 70121-78-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR