2-Bromo-4-fluoro-3-methyl-1-(methylsulfonyl)benzene ≥97%

Supplier: AOBChem USA

Warning

69003-500MG 69003-1G 69003-5G 69003-25G
77333-376EA 179.22 USD
77333-376 77346-744 77369-394 77390-412
2-Bromo-4-fluoro-3-methyl-1-(methylsulfonyl)benzene ≥97%
2-Bromo-4-fluoro-3-methyl-1-(methylsulfonyl)benzene

Precaución: for Research Use Only

Formula: C₈H₈BrFO₂S
MW: 267.115 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34763736
CAS Number: 2901098-24-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR