(2-Bromo-3-chloro-4-fluorophenyl)(methyl)sulfane ≥97%

Supplier: AOBChem USA

Warning

68999-1G 68999-5G 68999-25G
77341-644EA 97.85 USD
77341-644 77369-392 77381-490
(2-Bromo-3-chloro-4-fluorophenyl)(methyl)sulfane ≥97%
(2-Bromo-3-chloro-4-fluorophenyl)(methyl)sulfane

Precaución: for Research Use Only

Formula: C₇H₅BrClFS
MW: 255.535 g/mol
Storage Temperature: Ambient
CAS Number: 2706867-89-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR