(2-Bromo-4-fluoro-1,3-phenylene)bis(methylsulfane) ≥97%

Supplier: AOBChem USA

Warning

68998-500MG 68998-1G 68998-5G 68998-25G
77333-374EA 161.71 USD
77333-374 77345-980 77369-390 77390-410
(2-Bromo-4-fluoro-1,3-phenylene)bis(methylsulfane) ≥97%
(2-Bromo-4-fluoro-1,3-phenylene)bis(methylsulfane)

Precaución: for Research Use Only

Formula: C₈H₈BrFS₂
MW: 267.182 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34763732
CAS Number: 2807447-83-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR