(2-Bromo-4-fluoro-3-iodophenyl)(methyl)sulfane ≥97%

Supplier: AOBChem USA

Warning

68994-1G 68994-5G 68994-25G
77341-514EA 90.64 USD
77341-514 77369-388 77381-488
(2-Bromo-4-fluoro-3-iodophenyl)(methyl)sulfane ≥97%
(2-Bromo-4-fluoro-3-iodophenyl)(methyl)sulfane

Precaución: for Research Use Only

Formula: C₇H₅BrFIS
Storage Temperature: Ambient
MDL Number: MFCD34763731
CAS Number: 2709971-04-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR