2-Bromo-4-ethoxy-4'-(trifluoromethyl)-1,1'-biphenyl ≥97%

Supplier: AOBChem USA

Warning

68980-500MG 68980-1G 68980-5G
77339-394EA 584.01 USD
77339-394 77357-546 77381-486
2-Bromo-4-ethoxy-4'-(trifluoromethyl)-1,1'-biphenyl ≥97%
2-Bromo-4-ethoxy-4'-(trifluoromethyl)-1,1'-biphenyl

Precaución: for Research Use Only

Formula: C₁₅H₁₂BrF₃O
MW: 345.154 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34763720
CAS Number: 2901109-79-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR