2-Bromo-4-ethoxy-1,1'-biphenyl ≥97%

Supplier: AOBChem USA

Warning

68973-500MG 68973-1G 68973-5G
77339-516EA 594.31 USD
77339-516 77357-544 77381-484
2-Bromo-4-ethoxy-1,1'-biphenyl ≥97%
2-Bromo-4-ethoxy-1,1'-biphenyl

Precaución: for Research Use Only

Formula: C₁₄H₁₃BrO
MW: 277.156 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34763713
CAS Number: 2918879-13-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR