2-Bromo-4-fluoro-5-nitrobenzaldehyde ≥95%

Supplier: AOBChem USA

Warning

68971-500MG 68971-1G 68971-5G 68971-25G
77333-372EA 181.28 USD
77333-372 77346-870 77369-386 77390-408
2-Bromo-4-fluoro-5-nitrobenzaldehyde ≥95%
2-Bromo-4-fluoro-5-nitrobenzaldehyde

Precaución: for Research Use Only

Formula: C₇H₃BrFNO₃
Storage Temperature: Ambient
MDL Number: MFCD28739850
CAS Number: 1805556-76-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR