3-Bromo-5-fluoro-[1,1'-biphenyl]-4-amine ≥97%

Supplier: AOBChem USA

Warning

68966-500MG 68966-1G 68966-5G 68966-25G
77333-370EA 252.35 USD
77333-370 77357-542 77369-384 77390-406
3-Bromo-5-fluoro-[1,1'-biphenyl]-4-amine ≥97%
3-Bromo-5-fluoro-[1,1'-biphenyl]-4-amine

Precaución: for Research Use Only

Formula: C₁₂H₉BrFN
MW: 266.109 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34763712
CAS Number: 2807466-95-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR