3,4-Dibromo-2-fluoro-3'-methyl-1,1'-biphenyl ≥95%

Supplier: AOBChem USA

Warning

68960-500MG 68960-1G 68960-25G 68960-5G
77333-366EA 299.73 USD
77333-366 77357-538 77390-402 77381-482
3,4-Dibromo-2-fluoro-3'-methyl-1,1'-biphenyl ≥95%
3,4-Dibromo-2-fluoro-3'-methyl-1,1'-biphenyl

Precaución: for Research Use Only

Formula: C₁₃H₉Br₂F
MW: 344.017 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34763707
CAS Number: 2901083-35-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR