3,4-Dibromo-2-fluoro-1,1'-biphenyl ≥95%

Supplier: AOBChem USA

Warning

68957-500MG 68957-1G 68957-5G 68957-25G
77333-364EA 250.29 USD
77333-364 77357-536 77369-380 77390-400
3,4-Dibromo-2-fluoro-1,1'-biphenyl ≥95%
3,4-Dibromo-2-fluoro-1,1'-biphenyl

Precaución: for Research Use Only

Formula: C₁₂H₇Br₂F
Storage Temperature: Ambient
MDL Number: MFCD34763704
CAS Number: 2901102-68-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR