1,2,4-Tribromo-5-fluorobenzene ≥97%

Supplier: AOBChem USA

Warning

68943-500MG 68943-1G 68943-5G 68943-25G
77333-362EA 153.47 USD
77333-362 77345-520 77369-376 77390-398
1,2,4-Tribromo-5-fluorobenzene ≥97%
1,2,4-Tribromo-5-fluorobenzene

Precaución: for Research Use Only

Formula: C₆H₂Br₃F
MW: 332.79 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18089318
CAS Number: 7655-70-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR