Methyl-4-amino-3-iodo-5-methylbenzoate ≥97%

Supplier: AOBChem USA

Warning

Synonyms: Methyl 4-amino-3-iodo-5-methylbenzoate

68932-500MG 68932-1G 68932-5G 68932-25G
77333-360EA 112.27 USD
77333-360 77343-314 77369-374 77381-476
Methyl-4-amino-3-iodo-5-methylbenzoate ≥97%
Methyl-4-amino-3-iodo-5-methylbenzoate

Precaución: for Research Use Only

Formula: C₉H₁₀INO₂
MW: 291.09 g/mol
Storage Temperature: Ambient
MDL Number: MFCD10703532
CAS Number: 180624-11-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR