4,6-Dibromo-2-fluoro-3-methylbenzoic acid ≥97%

Supplier: AOBChem USA

Warning

68908-500MG 68908-1G 68908-5G 68908-25G
77333-358EA 258.53 USD
77333-358 77357-532 77369-372 77390-396
4,6-Dibromo-2-fluoro-3-methylbenzoic acid ≥97%
4,6-Dibromo-2-fluoro-3-methylbenzoic acid

Precaución: for Research Use Only

Formula: C₈H₅Br₂FO₂
Storage Temperature: Ambient
MDL Number: MFCD34763692
CAS Number: 2807445-37-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR