6-(2-Methoxyethoxy)-5-methylnicotinaldehyde ≥97%

Supplier: AOBChem USA

Warning

68902-500MG 68902-1G 68902-5G 68902-25G
77333-354EA 161.71 USD
77333-354 77345-978 77369-370 77390-392
6-(2-Methoxyethoxy)-5-methylnicotinaldehyde ≥97%
6-(2-Methoxyethoxy)-5-methylnicotinaldehyde

Precaución: for Research Use Only

Formula: C₁₀H₁₃NO₃
Storage Temperature: Ambient
CAS Number: 1515828-97-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR