2-(Cyclopropylmethoxy)-5-iodo-3-methylpyridine ≥97%

Supplier: AOBChem USA

Warning

68900-500MG 68900-1G 68900-5G
77336-558EA 388.31 USD
77336-558 77357-526 77381-470
2-(Cyclopropylmethoxy)-5-iodo-3-methylpyridine ≥97%
2-(Cyclopropylmethoxy)-5-iodo-3-methylpyridine

Precaución: for Research Use Only

Formula: C₁₀H₁₂INO
MW: 289.113 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34763687
CAS Number: 2807454-92-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR