5-(Methylthio)-2-(2,2,2-trifluoroethoxy)pyridine ≥97%

Supplier: AOBChem USA

Warning

68898-500MG 68898-1G 68898-5G 68898-25G
77333-352EA 141.11 USD
77333-352 77344-896 77369-368 77390-390
5-(Methylthio)-2-(2,2,2-trifluoroethoxy)pyridine ≥97%
5-(Methylthio)-2-(2,2,2-trifluoroethoxy)pyridine

Precaución: for Research Use Only

Formula: C₈H₈F₃NOS
MW: 223.215 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34763686
CAS Number: 2807465-62-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR