5-Iodo-2-(2,2,2-trifluoroethoxy)pyridine ≥95%

Supplier: AOBChem USA

Warning

68897-500MG 68897-1G 68897-5G 68897-25G
77333-350EA 116.39 USD
77333-350 77343-510 77369-366 77381-468
5-Iodo-2-(2,2,2-trifluoroethoxy)pyridine ≥95%
5-Iodo-2-(2,2,2-trifluoroethoxy)pyridine

Precaución: for Research Use Only

Formula: C₇H₅F₃INO
Storage Temperature: Ambient
MDL Number: MFCD18207566
CAS Number: 1252689-74-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR