3-Fluoro-2,5-dimethyl-4'-(trifluoromethyl)-1,1'-biphenyl

Supplier: AOBChem USA

Warning

68896-500MG 68896-1G 68896-25G 68896-5G
77333-348EA 285.31 USD
77333-348 77357-524 77390-388 77381-466
3-Fluoro-2,5-dimethyl-4'-(trifluoromethyl)-1,1'-biphenyl
3-Fluoro-2,5-dimethyl-4'-(trifluoromethyl)-1,1'-biphenyl

Precaución: for Research Use Only

Formula: C₁₅H₁₂F₄
MW: 268.249 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34763685
CAS Number: 2807471-90-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR