3-Fluoro-2,3',5-trimethyl-1,1'-biphenyl ≥95%

Supplier: AOBChem USA

Warning

68895-500MG 68895-1G 68895-5G
77333-346EA 320.33 USD
77333-346 77357-522 77381-464
3-Fluoro-2,3',5-trimethyl-1,1'-biphenyl ≥95%
3-Fluoro-2,3',5-trimethyl-1,1'-biphenyl

Precaución: for Research Use Only

Formula: C₁₅H₁₅F
MW: 214.278 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34763684
CAS Number: 2807442-86-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR