3-Fluoro-4'-methoxy-2,5-dimethyl-1,1'-biphenyl ≥97%

Supplier: AOBChem USA

Warning

68894-500MG 68894-1G 68894-5G
77333-344EA 320.33 USD
77333-344 77357-520 77381-462
3-Fluoro-4'-methoxy-2,5-dimethyl-1,1'-biphenyl ≥97%
3-Fluoro-4'-methoxy-2,5-dimethyl-1,1'-biphenyl

Precaución: for Research Use Only

Formula: C₁₅H₁₅FO
Storage Temperature: Ambient
MDL Number: MFCD34763683
CAS Number: 2807439-97-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR