3-Fluoro-2,4',5-trimethyl-1,1'-biphenyl ≥95%

Supplier: AOBChem USA

Warning

68893-500MG 68893-1G 68893-5G 68893-25G
77333-342EA 202.91 USD
77333-342 77357-518 77369-364 77390-386
3-Fluoro-2,4',5-trimethyl-1,1'-biphenyl ≥95%
3-Fluoro-2,4',5-trimethyl-1,1'-biphenyl

Precaución: for Research Use Only

Formula: C₁₅H₁₅F
MW: 214.278 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34763682
CAS Number: 2807443-65-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR