1-Fluoro-3-iodo-2,5-dimethylbenzene ≥97%

Supplier: AOBChem USA

Warning

68891-500MG 68891-1G 68891-5G 68891-25G
77333-338EA 250.29 USD
77333-338 77357-514 77369-362 77390-384
1-Fluoro-3-iodo-2,5-dimethylbenzene ≥97%
1-Fluoro-3-iodo-2,5-dimethylbenzene

Precaución: for Research Use Only

Formula: C₈H₈FI
Storage Temperature: Ambient
MDL Number: MFCD28794434
CAS Number: 1803815-93-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR