1-Chloro-3-fluoro-5-(methylsulfonyl)benzene ≥97%

Supplier: AOBChem USA

Warning

68886-500MG 68886-1G 68886-5G 68886-25G
77333-336EA 158.62 USD
77333-336 77345-742 77369-360 77390-382
1-Chloro-3-fluoro-5-(methylsulfonyl)benzene ≥97%
1-Chloro-3-fluoro-5-(methylsulfonyl)benzene

Precaución: for Research Use Only

Formula: C₇H₆ClFO₂S
MW: 208.638 g/mol
Storage Temperature: Ambient
MDL Number: MFCD25954296
CAS Number: 1020734-72-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR