(2-Chloro-6-fluoro-1,4-phenylene)bis(methylsulfane) ≥97%

Supplier: AOBChem USA

Warning

68882-500MG 68882-1G 68882-5G 68882-25G
77333-332EA 250.29 USD
77333-332 77357-508 77369-358 77390-380
(2-Chloro-6-fluoro-1,4-phenylene)bis(methylsulfane) ≥97%
(2-Chloro-6-fluoro-1,4-phenylene)bis(methylsulfane)

Precaución: for Research Use Only

Formula: C₈H₈ClFS₂
MW: 222.731 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34763678
CAS Number: 2901109-78-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR