2-Chloro-6-fluoro-4-(methylthio)benzoic acid ≥97%

Supplier: AOBChem USA

Warning

68880-500MG 68880-1G 68880-5G 68880-25G
77333-330EA 187.46 USD
77333-330 77357-506 77369-356 77390-378
2-Chloro-6-fluoro-4-(methylthio)benzoic acid ≥97%
2-Chloro-6-fluoro-4-(methylthio)benzoic acid

Precaución: for Research Use Only

Formula: C₈H₆ClFO₂S
MW: 220.648 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34763676
CAS Number: 2901081-91-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR