2-Fluoro-1,3-diiodobenzene ≥97%

Supplier: AOBChem USA

Warning

68876-500MG 68876-1G 68876-5G 68876-25G
77333-326EA 108.15 USD
77333-326 77343-206 77369-352 77381-454
2-Fluoro-1,3-diiodobenzene ≥97%
2-Fluoro-1,3-diiodobenzene

Precaución: for Research Use Only

Formula: C₆H₃FI₂
Storage Temperature: Ambient
MDL Number: MFCD28791294
CAS Number: 501433-04-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR