(3-Bromo-4-fluoro-5-methoxyphenyl)(methyl)sulfane ≥95%

Supplier: AOBChem USA

Warning

68875-500MG 68875-1G 68875-5G 68875-25G
77333-324EA 161.71 USD
77333-324 77345-976 77369-350 77390-374
(3-Bromo-4-fluoro-5-methoxyphenyl)(methyl)sulfane ≥95%
(3-Bromo-4-fluoro-5-methoxyphenyl)(methyl)sulfane

Precaución: for Research Use Only

Formula: C₈H₈BrFOS
MW: 251.116 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34763673
CAS Number: 2710912-81-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR