2,4-Difluoro-3-(trimethylsilyl)benzaldehyde ≥97%

Supplier: AOBChem USA

Warning

68874-500MG 68874-1G 68874-5G
77337-800EA 444.96 USD
77337-800 77357-502 77381-452
2,4-Difluoro-3-(trimethylsilyl)benzaldehyde ≥97%
2,4-Difluoro-3-(trimethylsilyl)benzaldehyde

Precaución: for Research Use Only

Formula: C₁₀H₁₂F₂OSi
MW: 214.284 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34763672
CAS Number: 2901095-75-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR