(2,4-Difluoro-3-(trimethylsilyl)phenyl)boronic acid ≥97%

Supplier: AOBChem USA

Warning

68872-500MG 68872-1G 68872-5G
77333-320EA 337.84 USD
77333-320 77357-498 77381-448
(2,4-Difluoro-3-(trimethylsilyl)phenyl)boronic acid ≥97%
(2,4-Difluoro-3-(trimethylsilyl)phenyl)boronic acid

Precaución: for Research Use Only

Formula: C₉H₁₃BF₂O₂Si
MW: 230.092 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18384122
CAS Number: 1190989-14-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR