4-Chlorobenzo[d][1,3]dioxole-5-carboxylic acid ≥97%

Supplier: AOBChem USA

Warning

68850-500MG 68850-1G 68850-5G
77337-678EA 435.69 USD
77337-678 77357-492 77381-442
4-Chlorobenzo[d][1,3]dioxole-5-carboxylic acid ≥97%
4-Chlorobenzo[d][1,3]dioxole-5-carboxylic acid

Precaución: for Research Use Only

Formula: C₈H₅ClO₄
Storage Temperature: Ambient
MDL Number: MFCD34763668
CAS Number: 890927-00-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR