(4-Chlorobenzo[d][1,3]dioxol-5-yl)boronic acid ≥95%

Supplier: AOBChem USA

Warning

68848-500MG 68848-1G 68848-5G
77339-514EA 594.31 USD
77339-514 77357-490 77381-440
(4-Chlorobenzo[d][1,3]dioxol-5-yl)boronic acid ≥95%
(4-Chlorobenzo[d][1,3]dioxol-5-yl)boronic acid

Precaución: for Research Use Only

Formula: C₇H₆BClO₄
Storage Temperature: Ambient
MDL Number: MFCD23380968
CAS Number: 1392415-62-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR