(2-(Benzyloxy)phenyl)trimethylsilane ≥95%

Supplier: AOBChem USA

Warning

68840-1G 68840-5G 68840-25G
77341-364EA 82.4 USD
77341-364 77369-346 77381-436
(2-(Benzyloxy)phenyl)trimethylsilane ≥95%
(2-(Benzyloxy)phenyl)trimethylsilane

Precaución: for Research Use Only

Formula: C₁₆H₂₀OSi
MW: 256.415 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34763665
CAS Number: 2807456-30-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR