(3,5-Dimethylphenyl)trimethylsilane ≥97%

Supplier: AOBChem USA

Warning

68838-500MG 68838-1G 68838-5G 68838-25G
77333-318EA 220.42 USD
77333-318 77357-488 77369-342 77390-370
(3,5-Dimethylphenyl)trimethylsilane ≥97%
(3,5-Dimethylphenyl)trimethylsilane

Precaución: for Research Use Only

Formula: C₁₁H₁₈Si
MW: 178.346 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06201832
CAS Number: 17961-83-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR