(2,5-Dimethylphenyl)trimethylsilane 95

Supplier: AOBChem USA

Warning

68834-500MG 68834-1G 68834-5G 68834-25G
77333-314EA 161.71 USD
77333-314 77345-974 77369-338 77390-366
(2,5-Dimethylphenyl)trimethylsilane 95
(2,5-Dimethylphenyl)trimethylsilane

Precaución: for Research Use Only

Formula: C₁₁H₁₈Si
MW: 178.346 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34687422
CAS Number: 17961-81-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR