3-(Trimethylsilyl)anisole ≥97%

Supplier: AOBChem USA

Warning

Synonyms: (3-Methoxyphenyl)trimethylsilane

68828-1G 68828-5G 68828-25G
77341-058EA 67.98 USD
77341-058 77369-330 77381-426
3-(Trimethylsilyl)anisole ≥97%
3-(Trimethylsilyl)anisole

Precaución: for Research Use Only

Formula: C₁₀H₁₆OSi
MW: 180.319 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06201812
CAS Number: 17876-90-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR