2,4-Difluoro-3-iodo-4'-methoxy-1,1'-biphenyl ≥97%

Supplier: AOBChem USA

Warning

68823-500MG 68823-1G 68823-5G 68823-25G
77333-308EA 168.92 USD
77333-308 77346-308 77369-328 77390-364
2,4-Difluoro-3-iodo-4'-methoxy-1,1'-biphenyl ≥97%
2,4-Difluoro-3-iodo-4'-methoxy-1,1'-biphenyl

Precaución: for Research Use Only

Formula: C₁₃H₉F₂IO
MW: 346.111 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34763660
CAS Number: 2901102-21-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR