2,4-Difluoro-4'-methoxy-[1,1'-biphenyl]-3-carboxylic acid ≥97%

Supplier: AOBChem USA

Warning

68819-500MG 68819-1G 68819-5G 68819-25G
77333-304EA 168.92 USD
77333-304 77346-306 77369-324 77390-360
2,4-Difluoro-4'-methoxy-[1,1'-biphenyl]-3-carboxylic acid ≥97%
2,4-Difluoro-4'-methoxy-[1,1'-biphenyl]-3-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₄H₁₀F₂O₃
MW: 264.224 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34763656
CAS Number: 2901095-31-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR