2,2-Difluoro-1-(5-(methylthio)thiophen-2-yl)ethanone ≥97%

Supplier: AOBChem USA

Warning

68815-500MG 68815-1G 68815-5G 68815-25G
77333-300EA 116.39 USD
77333-300 77343-506 77369-320 77381-424
2,2-Difluoro-1-(5-(methylthio)thiophen-2-yl)ethanone ≥97%
2,2-Difluoro-1-(5-(methylthio)thiophen-2-yl)ethanone

Precaución: for Research Use Only

Formula: C₇H₆F₂OS₂
MW: 208.249 g/mol
Storage Temperature: Ambient
CAS Number: 2901083-29-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR